4-(Hydroxymethyl)-1,3-oxazole
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(1,3-Oxazol-4-yl)methanol, 4-(Hydroxymethyl)oxazole
OR59847-1G
OR59847-250MG
APOSOR59847-1GEA
109
EUR
APOSOR59847-1G
APOSOR59847-250MG
4-(Hydroxymethyl)-1,3-oxazole
4-(Hydroxymethyl)-1,3-oxazole
Formel:
C₄H₅NO₂ Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD08703646 CAS-Nummer: 155742-48-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...