5-Carboxy-2-chloro-4-methoxybenzeneboronic acid
Lieferant: Apollo Scientific
Synonyme:
5-Borono-4-chloro-2-methoxybenzoic acid, 5-Carboxy-2-chloro-4-methoxyphenylboronic acid
OR59414-1G
OR59414-250MG
APOSOR59414-1GEA
437
EUR
APOSOR59414-1G
APOSOR59414-250MG
5-Carboxy-2-chloro-4-methoxybenzeneboronic acid
5-Carboxy-2-chloro-4-methoxybenzeneboronic acid
Formel:
C₈H₈BClO₅ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD09878319 CAS-Nummer: 957066-09-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...