5-Bromo-4-chloro-2-methoxybenzoic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromo-2-carboxy-5-chloroanisole, 5-Bromo-4-chloro-o-anisic acid

OR59335-1G OR59335-5G OR59335-250MG
APOSOR59335-1GEA 141 EUR
APOSOR59335-1G APOSOR59335-5G APOSOR59335-250MG
5-Bromo-4-chloro-2-methoxybenzoic acid
5-Bromo-4-chloro-2-methoxybenzoic acid
Formel: C₈H₆BrClO₃
Molekulargewicht: 265.488 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD09878363
CAS-Nummer: 95383-17-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR