Diethyl 4-iodopyridine-2,6-dicarboxylate 98+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Iodopyridine-2,6-dicarboxylic acid diethyl ester

OR55111-1G OR55111-250MG
APOSOR55111-1GEA 382 EUR
APOSOR55111-1G APOSOR55111-250MG
Diethyl 4-iodopyridine-2,6-dicarboxylate 98+%
Diethyl 4-iodopyridine-2,6-dicarboxylate
Formel: C₁₁H₁₂INO₄
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD11042539
CAS-Nummer: 120491-90-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR