3-Bromo-5-(methylsulphonyl)aniline 98%

Lieferant: Apollo Scientific

OR55091-250MG OR55091-1G
APOSOR55091-250MGEA 105 EUR
APOSOR55091-250MG APOSOR55091-1G
3-Bromo-5-(methylsulphonyl)aniline 98%
3-Bromo-5-(methylsulphonyl)aniline
Formel: C₇H₈BrNO₂S
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD17392822
CAS-Nummer: 62606-00-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR