cis-3-Aminocyclohexanecarboxylic acid, N-FMOC protected

Lieferant: Apollo Scientific
Achtung

Synonyme: cis-3-{[(9H-Fluoren-9-ylmethoxy)carbonyl]amino}cyclohexanecarboxylic acid

OR53198-1G OR53198-5G
APOSOR53198-1GEA 219 EUR
APOSOR53198-1G APOSOR53198-5G
cis-3-Aminocyclohexanecarboxylic acid, N-FMOC protected
cis-3-Aminocyclohexanecarboxylic acid, N-FMOC protected
Formel: C₂₂H₂₃NO₄
Molekulargewicht: 365.422 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD01862345
CAS-Nummer: 312965-05-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR