5-Chloro-1H-pyrazolo[3,4-b]pyridin-3-amine

Lieferant: Apollo Scientific
Achtung

OR52980-1G OR52980-250MG
APOSOR52980-1GEA 383 EUR
APOSOR52980-1G APOSOR52980-250MG
5-Chloro-1H-pyrazolo[3,4-b]pyridin-3-amine
5-Chloro-1H-pyrazolo[3,4-b]pyridin-3-amine
Formel: C₆H₅ClN₄
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD14706816
CAS-Nummer: 1245643-75-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR