2-Chloro-N,N-diisobutylacetamide

Lieferant: Apollo Scientific
Achtung

OR52317-10G OR52317-1G OR52317-5G
APOSOR52317-10GEA 184 EUR
APOSOR52317-10G APOSOR52317-1G APOSOR52317-5G
2-Chloro-N,N-diisobutylacetamide
2-Chloro-N,N-diisobutylacetamide
Formel: C₁₀H₂₀ClNO
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00040035
CAS-Nummer: 5326-82-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR