Methyl 4,5,6,7-tetrahydrobenzo[b]thiophene-2-carboxylate

Lieferant: Apollo Scientific

Synonyme: 2-(Methoxycarbonyl)-4,5,6,7-tetrahydrobenzo[b]thiophene, Methyl 4,5,6,7-tetrahydro-1-benzothiophene-2-carboxylate

OR52032-10G OR52032-1G OR52032-5G
APOSOR52032-10GEA 799 EUR
APOSOR52032-10G APOSOR52032-1G APOSOR52032-5G
Methyl 4,5,6,7-tetrahydrobenzo[b]thiophene-2-carboxylate
Methyl 4,5,6,7-tetrahydrobenzo[b]thiophene-2-carboxylate
Formel: C₁₀H₁₂O₂S
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD06654963
CAS-Nummer: 91489-09-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR