(3-Methoxyphenyl)methanethiol 95%
Lieferant: Apollo Scientific
OR51960-1G
OR51960-250MG
APOSOR51960-1GEA
719
EUR
APOSOR51960-1G
APOSOR51960-250MG
(3-Methoxyphenyl)methanethiol 95%
(3-Methoxyphenyl)methanethiol
Formel:
C₈H₁₀OS Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD09812092 CAS-Nummer: 7166-64-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...