4-Acetyl-2-bromobenzoic acid

Lieferant: Apollo Scientific
Achtung

OR50625-1G OR50625-250MG
APOSOR50625-1GEA 654 EUR
APOSOR50625-1G APOSOR50625-250MG
4-Acetyl-2-bromobenzoic acid
4-Acetyl-2-bromobenzoic acid
Formel: C₉H₇BrO₃
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD26520854
CAS-Nummer: 93273-64-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR