2-Bromo-1-(4-chloro-1-methyl-1H-pyrazol-3-yl)ethanone 97%

Lieferant: Apollo Scientific
Achtung

OR500028-1G OR500028-250MG
APOSOR500028-1GEA 328 EUR
APOSOR500028-1G APOSOR500028-250MG
2-Bromo-1-(4-chloro-1-methyl-1H-pyrazol-3-yl)ethanone 97%
2-Bromo-1-(4-chloro-1-methyl-1H-pyrazol-3-yl)ethanone
Formel: C₆H₆BrClN₂O
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD30502621
CAS-Nummer: 2068151-92-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR