6-Chloroquinolin-8-amine 95+%

Lieferant: Apollo Scientific
Achtung

OR500021-1G OR500021-250MG
APOSOR500021-1GEA 101 EUR
APOSOR500021-1G APOSOR500021-250MG
6-Chloroquinolin-8-amine 95+%
6-Chloroquinolin-8-amine
Formel: C₉H₇ClN₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD09049101
CAS-Nummer: 5470-75-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR