3-Bromo-4-(hydroxymethyl)phenol 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR500011-100MG
OR500011-250MG
OR500011-1G
APOSOR500011-100MGEA
84.1
EUR
APOSOR500011-100MG
APOSOR500011-250MG
APOSOR500011-1G
3-Bromo-4-(hydroxymethyl)phenol 97%
3-Bromo-4-(hydroxymethyl)phenol
Formel:
C₇H₇BrO₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD18391433 CAS-Nummer: 666747-04-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...