4-Chloro-3-(hydroxymethyl)phenol 95+%
Lieferant: Apollo Scientific
|
Achtung
|
OR500010-1G
OR500010-250MG
APOSOR500010-1GEA
84.1
EUR
APOSOR500010-1G
APOSOR500010-250MG
4-Chloro-3-(hydroxymethyl)phenol 95+%
4-Chloro-3-(hydroxymethyl)phenol
Formel:
C₇H₇ClO₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD09038291 CAS-Nummer: 876299-47-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...