4-Bromo-3-iodonitrobenzene
Lieferant: Apollo Scientific
|
Achtung
|
OR48259-1G
OR48259-5G
OR48259-25G
APOSOR48259-1GEA
62.5
EUR
APOSOR48259-1G
APOSOR48259-5G
APOSOR48259-25G
4-Bromo-3-iodonitrobenzene
Formel:
C₆H₃BrINO₂ Schmelzpunkt: 94…99 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD17012734 CAS-Nummer: 63037-63-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...