5-Bromo-2-cyclobutoxypyrimidine 95%
Lieferant: Apollo Scientific
OR471698-1G
OR471698-250MG
OR471698-5G
APOSOR471698-1GEA
652
EUR
APOSOR471698-1G
APOSOR471698-250MG
APOSOR471698-5G
5-Bromo-2-cyclobutoxypyrimidine 95%
5-Bromo-2-cyclobutoxypyrimidine
Formel:
C₈H₉BrN₂O Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD16618818 CAS-Nummer: 1260833-40-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...