Methyl 5-chloroindole-3-carboxylate 97%

Lieferant: Apollo Scientific

Synonyme: Methyl 5-chloro-1H-indole-3-carboxylate

OR471689-1G OR471689-250MG OR471689-5G
APOSOR471689-1GEA 58 EUR
APOSOR471689-1G APOSOR471689-250MG APOSOR471689-5G
Methyl 5-chloroindole-3-carboxylate 97%
Methyl 5-chloroindole-3-carboxylate
Formel: C₁₀H₈ClNO₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD06203871
CAS-Nummer: 172595-67-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR