1-Benzyl-4-methyl-3-(methylamino)piperidine dihydrochloride 97%

Lieferant: Apollo Scientific

OR471680-10G OR471680-1G OR471680-5G
APOSOR471680-10GEA 102 EUR
APOSOR471680-10G APOSOR471680-1G APOSOR471680-5G
1-Benzyl-4-methyl-3-(methylamino)piperidine dihydrochloride 97%
1-Benzyl-4-methyl-3-(methylamino)piperidine dihydrochloride
Formel: C₁₄H₂₂N₂·2ClH
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD09475546
CAS-Nummer: 1228879-37-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR