5-Chloro-2,4-dimethoxypyrimidine
Lieferant: Apollo Scientific
OR471528-5G
OR471528-1G
OR471528-250MG
APOSOR471528-5GEA
364
EUR
APOSOR471528-5G
APOSOR471528-1G
APOSOR471528-250MG
5-Chloro-2,4-dimethoxypyrimidine
5-Chloro-2,4-dimethoxypyrimidine
Formel:
C₆H₇ClN₂O₂ Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD13619877 CAS-Nummer: 123551-49-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...