Tetrakis(4-cyanophenyl)methane

Lieferant: Apollo Scientific

OR471492-1G OR471492-250MG
APOSOR471492-1GEA 188 EUR
APOSOR471492-1G APOSOR471492-250MG
Tetrakis(4-cyanophenyl)methane
Tetrakis(4-cyanophenyl)methane
Formel: C₂₉H₁₆N₄
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD29049230
CAS-Nummer: 121706-21-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR