α-Cyclobutylbenzylamine hydrochloride

Lieferant: Apollo Scientific

Synonyme: Cyclobutyl(phenyl)methanamine hydrochloride

OR471346-1G OR471346-250MG
APOSOR471346-1GEA 447 EUR
APOSOR471346-1G APOSOR471346-250MG
α-Cyclobutylbenzylamine hydrochloride
α-Cyclobutylbenzylamine hydrochloride
Formel: C₁₁H₁₅N·ClH
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD28126674
CAS-Nummer: 58271-61-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR