5-Bromo-2-methylquinoline
Lieferant: Apollo Scientific
OR470776-1G
OR470776-250MG
OR470776-5G
APOSOR470776-1GEA
257
EUR
APOSOR470776-1G
APOSOR470776-250MG
APOSOR470776-5G
5-Bromo-2-methylquinoline
Formel:
C₁₀H₈BrN Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD13189490 CAS-Nummer: 54408-52-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...