2-Amino-2-(3-thienyl)acetic acid hydrochloride

Lieferant: Apollo Scientific

OR470332-5G OR470332-1G
APOSOR470332-5GEA 1540 EUR
APOSOR470332-5G APOSOR470332-1G
2-Amino-2-(3-thienyl)acetic acid hydrochloride
2-Amino-2-(3-thienyl)acetic acid hydrochloride
Formel: C₆H₇NO₂S·ClH
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD02683175
CAS-Nummer: 369403-64-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR