2-Chloro-4-(methylsulphonamido)benzoic acid
Lieferant: Apollo Scientific
OR470320-1G
OR470320-5G
APOSOR470320-1GEA
245
EUR
APOSOR470320-1G
APOSOR470320-5G
2-Chloro-4-(methylsulphonamido)benzoic acid
2-Chloro-4-(methylsulphonamido)benzoic acid
Formel:
C₈H₈ClNO₄S Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD14659025 CAS-Nummer: 158579-73-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...