5-Hydroxy-2-(methylsulphonamido)benzoic acid
Lieferant: Apollo Scientific
OR470319-1G
OR470319-5G
APOSOR470319-1GEA
275
EUR
APOSOR470319-1G
APOSOR470319-5G
5-Hydroxy-2-(methylsulphonamido)benzoic acid
5-Hydroxy-2-(methylsulphonamido)benzoic acid
Formel:
C₈H₉NO₅S Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD14660388 CAS-Nummer: 1243391-74-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...