5-Bromo-2-(methylsulphonamido)benzoic acid
Lieferant: Apollo Scientific
OR470317-1G
OR470317-5G
APOSOR470317-1GEA
245
EUR
APOSOR470317-1G
APOSOR470317-5G
5-Bromo-2-(methylsulphonamido)benzoic acid
5-Bromo-2-(methylsulphonamido)benzoic acid
Formel:
C₈H₈BrNO₄S Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00452520 CAS-Nummer: 1225713-29-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...