2-Chloro-6-[(cyclopropylcarbonyl)amino]benzoic acid
Lieferant: Apollo Scientific
OR470312-1G
OR470312-5G
APOSOR470312-1GEA
245
EUR
APOSOR470312-1G
APOSOR470312-5G
2-Chloro-6-[(cyclopropylcarbonyl)amino]benzoic acid
2-Chloro-6-[(cyclopropylcarbonyl)amino]benzoic acid
Formel:
C₁₁H₁₀ClNO₃ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD18872285 CAS-Nummer: 1314406-53-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...