3-[(Cyclopropylcarbonyl)amino]-2-thiophenecarboxylic acid
Lieferant: Apollo Scientific
OR470310-1G
OR470310-5G
APOSOR470310-1GEA
275
EUR
APOSOR470310-1G
APOSOR470310-5G
3-[(Cyclopropylcarbonyl)amino]-2-thiophenecarboxylic acid
3-[(Cyclopropylcarbonyl)amino]-2-thiophenecarboxylic acid
Formel:
C₉H₉NO₃S Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD09047601 CAS-Nummer: 926273-18-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...