2-Chloro-4-[(cyclopropylcarbonyl)amino]benzoic acid
Lieferant: Apollo Scientific
OR470308-1G
OR470308-5G
APOSOR470308-1GEA
245
EUR
APOSOR470308-1G
APOSOR470308-5G
2-Chloro-4-[(cyclopropylcarbonyl)amino]benzoic acid
2-Chloro-4-[(cyclopropylcarbonyl)amino]benzoic acid
Formel:
C₁₁H₁₀ClNO₃ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD14659014 CAS-Nummer: 1249352-65-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...