2-[(Cyclopropylcarbonyl)amino]-5-hydroxybenzoic acid

Lieferant: Apollo Scientific

OR470306-1G OR470306-5G
APOSOR470306-1GEA 275 EUR
APOSOR470306-1G APOSOR470306-5G
2-[(Cyclopropylcarbonyl)amino]-5-hydroxybenzoic acid
2-[(Cyclopropylcarbonyl)amino]-5-hydroxybenzoic acid
Formel: C₁₁H₁₁NO₄
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD03081976
CAS-Nummer: 543695-30-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR