2-(2-Naphthyloxy)benzonitrile

Lieferant: Apollo Scientific

OR470251-1G OR470251-5G
APOSOR470251-1GEA 164 EUR
APOSOR470251-1G APOSOR470251-5G
2-(2-Naphthyloxy)benzonitrile
Formel: C₁₇H₁₁NO
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD12618618
CAS-Nummer: 1041593-26-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR