2-Amino-5-(4-methyl-1-piperazinyl)benzoic acid

Lieferant: Apollo Scientific

OR470074-1G OR470074-5G OR470074-250MG
APOSOR470074-1GEA 463 EUR
APOSOR470074-1G APOSOR470074-5G APOSOR470074-250MG
2-Amino-5-(4-methyl-1-piperazinyl)benzoic acid
2-Amino-5-(4-methyl-1-piperazinyl)benzoic acid
Formel: C₁₂H₁₇N₃O₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD12869794
CAS-Nummer: 835879-06-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR