L-Tryptophan benzyl ester
Lieferant: Apollo Scientific
|
Achtung
|
OR46743-1G
OR46743-5G
APOSOR46743-1GEA
50.9
EUR
APOSOR46743-1G
APOSOR46743-5G
L-Tryptophan benzyl ester
L-Tryptophan benzyl ester
Formel:
C₁₈H₁₈N₂O₂ Schmelzpunkt: 77…80 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD02683457 CAS-Nummer: 4299-69-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...