2-(1-(Hydroxymethyl)cyclopropyl)acetic acid

Lieferant: Apollo Scientific

OR46663-1G OR46663-50MG OR46663-250MG OR46663-500MG OR46663-5G
APOSOR46663-1GEA 1040 EUR
APOSOR46663-1G APOSOR46663-50MG APOSOR46663-250MG APOSOR46663-500MG APOSOR46663-5G
2-(1-(Hydroxymethyl)cyclopropyl)acetic acid
2-(1-(Hydroxymethyl)cyclopropyl)acetic acid
Formel: C₆H₁₀O₃
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD09833495
CAS-Nummer: 142148-13-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR