5-Bromo-N-cyclopropylpyridin-2-amine 98%
Lieferant: Apollo Scientific
Synonyme:
N-(5-Bromopyridin-2-yl)cyclopropylamine, 5-Bromo-2-(cyclopropylamino)pyridine
OR46049-1G
OR46049-5G
APOSOR46049-1GEA
140
EUR
APOSOR46049-1G
APOSOR46049-5G
5-Bromo-N-cyclopropylpyridin-2-amine 98%
5-Bromo-N-cyclopropylpyridin-2-amine
Formel:
C₈H₉BrN₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD08706291 CAS-Nummer: 885266-96-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...