2-Nitro-phenethylamine hydrochloride 97%

Lieferant: Apollo Scientific

OR452217-1G OR452217-5G
APOSOR452217-1GEA 314 EUR
APOSOR452217-1G APOSOR452217-5G
2-Nitro-phenethylamine hydrochloride 97%
2-Nitro-phenethylamine hydrochloride
Formel: C₈H₁₀N₂O₂·ClH
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD07371521
CAS-Nummer: 861337-74-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR