N-Boc-[(S)-1-(aminomethyl)-2-methylpropyl]amine 97%

Lieferant: Apollo Scientific

OR452202-1G OR452202-250MG OR452202-500MG
APOSOR452202-1GEA 870 EUR
APOSOR452202-1G APOSOR452202-250MG APOSOR452202-500MG
N-Boc-[(S)-1-(aminomethyl)-2-methylpropyl]amine 97%
N-Boc-[(S)-1-(aminomethyl)-2-methylpropyl]amine
Formel: C₁₀H₂₂N₂O₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD08726033
CAS-Nummer: 400652-49-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR