2-Acetyl-4-(cyclopropyl)thiophene

Lieferant: Apollo Scientific

OR45218-1G OR45218-250MG
APOSOR45218-1GEA 654 EUR
APOSOR45218-1G APOSOR45218-250MG
2-Acetyl-4-(cyclopropyl)thiophene
2-Acetyl-4-(cyclopropyl)thiophene
Formel: C₉H₁₀OS
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD22068104
CAS-Nummer: 1021432-58-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR