(R)-a-Phenyl-1-pyrrolidineethanamine dihydrochloride 97%

Lieferant: Apollo Scientific

OR452156-1G OR452156-250MG OR452156-500MG
APOSOR452156-1GEA 959 EUR
APOSOR452156-1G APOSOR452156-250MG APOSOR452156-500MG
(R)-a-Phenyl-1-pyrrolidineethanamine dihydrochloride 97%
(R)-a-Phenyl-1-pyrrolidineethanamine dihydrochloride
Formel: C₁₂H₁₈N₂·2ClH
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD31560200
CAS-Nummer: 180305-85-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR