Boc-DL-norleucine 97%
Lieferant: Apollo Scientific
OR452085-1G
OR452085-25G
OR452085-5G
APOSOR452085-1GEA
109
EUR
APOSOR452085-1G
APOSOR452085-25G
APOSOR452085-5G
Boc-DL-norleucine 97%
Boc-DL-norleucine
Formel:
C₁₁H₂₁NO₄ Molekulargewicht: 231.289 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD01326593 CAS-Nummer: 125342-48-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...