1-Boc-3-azido-azetidine 97%

Lieferant: Apollo Scientific

OR452073-1G OR452073-250MG OR452073-500MG
APOSOR452073-1GEA 654 EUR
APOSOR452073-1G APOSOR452073-250MG APOSOR452073-500MG
1-Boc-3-azido-azetidine 97%
1-Boc-3-azido-azetidine
Formel: C₈H₁₄N₄O₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD19686277
CAS-Nummer: 429672-02-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR