4-Methyl-L-tryptophan 97%

Lieferant: Apollo Scientific

OR452025-250MG OR452025-500MG OR452025-100MG OR452025-1G
APOSOR452025-250MGEA 328 EUR
APOSOR452025-250MG APOSOR452025-500MG APOSOR452025-100MG APOSOR452025-1G
4-Methyl-L-tryptophan 97%
4-Methyl-L-tryptophan
Formel: C₁₂H₁₄N₂O₂
Molekulargewicht: 218.252 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD06797707
CAS-Nummer: 33468-33-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR