3-Cyclohexyl-DL-alanine 97%
Lieferant: Apollo Scientific
Synonyme:
2-Amino-3-cyclohexylpropanoic acid
OR452018-1G
OR452018-5G
APOSOR452018-1GEA
31.9
EUR
APOSOR452018-1G
APOSOR452018-5G
3-Cyclohexyl-DL-alanine 97%
3-Cyclohexyl-DL-alanine
Formel:
C₉H₁₇NO₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00996128 CAS-Nummer: 4441-50-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...