2-Bromo-4-iodo-6-methylphenol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Bromo-4-iodo-o-cresol, 3-Bromo-2-hydroxy-5-iodotoluene
OR45128-1G
OR45128-250MG
OR45128-5G
APOSOR45128-1GEA
227
EUR
APOSOR45128-1G
APOSOR45128-250MG
APOSOR45128-5G
2-Bromo-4-iodo-6-methylphenol
2-Bromo-4-iodo-6-methylphenol
Formel:
C₇H₆BrIO Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD27664804 CAS-Nummer: 1820641-98-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...