4-({[1-(tert-Butoxycarbonyl)piperidin-4-yl]oxy}methyl)benzaldehyde 90%

Lieferant: Apollo Scientific

Synonyme: 1-[(4-Formylbenzyl)oxy]piperidine, N-BOC protected

OR45070-1G OR45070-500MG
APOSOR45070-1GEA 236 EUR
APOSOR45070-1G APOSOR45070-500MG
4-({[1-(tert-Butoxycarbonyl)piperidin-4-yl]oxy}methyl)benzaldehyde 90%
4-({[1-(tert-Butoxycarbonyl)piperidin-4-yl]oxy}methyl)benzaldehyde
Formel: C₁₈H₂₅NO₄
Schmelzpunkt: 65.5…67 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD11841070
CAS-Nummer: 1080028-74-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR