Cyclohexanecarbonyl isocyanate

Lieferant: Apollo Scientific

OR450130-1G OR450130-250MG
APOSOR450130-1GEA 429 EUR
APOSOR450130-1G APOSOR450130-250MG
Cyclohexanecarbonyl isocyanate
Cyclohexanecarbonyl isocyanate

May form small amount of sediment

Formel: C₈H₁₁NO₂
Lagerungstemperatur: Gefrierschrank
MDL Number: MFCD16300873
CAS-Nummer: 4461-26-1
UN-Nummer: 2206
ADR: 6.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR