7-Amino-[1,2,4]triazolo[1,5-a]pyrimidine-6-carbonitrile
Lieferant: Apollo Scientific
OR450089-1G
OR450089-250MG
APOSOR450089-1GEA
161
EUR
APOSOR450089-1G
APOSOR450089-250MG
7-Amino-[1,2,4]triazolo[1,5-a]pyrimidine-6-carbonitrile
7-Amino-[1,2,4]triazolo[1,5-a]pyrimidine-6-carbonitrile
Formel:
C₆H₄N₆ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD07657833 CAS-Nummer: 28524-64-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...