3-(Cyclopropyl)thiophene
Lieferant: Apollo Scientific
|
Achtung
|
OR40807-1G
OR40807-250MG
APOSOR40807-1GEA
77
EUR
APOSOR40807-1G
APOSOR40807-250MG
3-(Cyclopropyl)thiophene
3-(Cyclopropyl)thiophene
Formel:
C₇H₈S Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD25962002 CAS-Nummer: 29576-51-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...