Methyl 2-chloro-5-methoxybenzoate 98%

Lieferant: Apollo Scientific
Achtung

OR400977-1G OR400977-5G
APOSOR400977-1GEA 59.6 EUR
APOSOR400977-1G APOSOR400977-5G
Methyl 2-chloro-5-methoxybenzoate 98%
Methyl 2-chloro-5-methoxybenzoate
Formel: C₉H₉ClO₃
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD12913365
CAS-Nummer: 54810-63-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR